Learn More
TARGETMOL CHEMICALS INC BIMU 8 5MG
SDP
Catalog No. 502580990
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BIMU 8 is a selective agonist of 5-HT4 with EC50s of 18 nM 77 nM and 540 nM for wild-type 5HT4 receptor T3.36A and W6.48A mutant 5-HT4. purity: 99%
Catalog No. 50-258-0990 Supplier TARGETMOL CHEMICALS INC Supplier No. T219465MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BIMU 8 is a selective agonist of 5-HT4 with EC50s of 18 nM 77 nM and 540 nM for wild-type 5HT4 receptor T3.36A and W6.48A mutant 5-HT4. purity: 99%