Learn More
MIDWEST WHOLESALE CONTAINER AkroBin 14 3/4" x 5 1/2" x 5"
Catalog No. NC1444787
Encompass_Preferred
This item has a minimum order quantity of 12 per supplier requirements. Optimize your storage space. Heavy-duty, polypropylene bins stack together or hang from Akro-Mils louvered racks and panels, rail racks, and carts for high-density storage. AkroBins are corrosion-proof and unaffected by weak acids and alkalis.
Catalog No. NC1444787 Supplier MIDWEST WHOLESALE CONTAINER Supplier No. 30234SCLAR
This item has a minimum order quantity of 12 per supplier requirements. Optimize your storage space. Heavy-duty, polypropylene bins stack together or hang from Akro-Mils louvered racks and panels, rail racks, and carts for high-density storage. AkroBins are corrosion-proof and unaffected by weak acids and alkalis.