Learn More
Med Vet International Dispensing Bin, Clear, Small, 5 1/2? x 9 1/2? x 9?, J1153, Each
Catalog No. 5000495957
Encompass_Preferred
Organize and upgrade your work area, retail space, wherever you need quick access to small items. These attractive dispensing bins have a magnet on the hopper door to keep it open during work periods. Hinged top door for convenient filling. Contents are visible yet kept dust free. Wall-mountable with hardware included. Available with bracket for slatwall attachment. Made of sparkling clear acrylic. Anti-skid rubber feet. Made in USA.These attractive storage bins are great for keeping high use items close at hand.• Three different sizes• Clear rigged polycarbonate: can easily see contents • Lid has magnetic back so can stay openSome suggested uses• Dispensing vials • Sampling jars• Blood tubesClear dispensing bin small 5 1/2” w x 9 1/2” x 9”h
Catalog No. 50-004-95957 Supplier Med Vet International Supplier No. JOR1153
Organize and upgrade your work area, retail space, wherever you need quick access to small items. These attractive dispensing bins have a magnet on the hopper door to keep it open during work periods. Hinged top door for convenient filling. Contents are visible yet kept dust free. Wall-mountable with hardware included. Available with bracket for slatwall attachment. Made of sparkling clear acrylic. Anti-skid rubber feet. Made in USA.These attractive storage bins are great for keeping high use items close at hand.• Three different sizes• Clear rigged polycarbonate: can easily see contents • Lid has magnetic back so can stay openSome suggested uses• Dispensing vials • Sampling jars• Blood tubesClear dispensing bin small 5 1/2” w x 9 1/2” x 9”h