Learn More
Aviva Systems Biology Corp BIN2 Antibody - middle region (ARP56880_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56880_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BIN2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human BIN2. Alternate names: BRAP-1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-286-229