Learn More
Aviva Systems Biology Corp BIN3 Antibody - C-terminal region (ARP65113_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65113_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BIN3. It was validated on Western Blot. . Alternate names: MGC14978ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-433