Learn More
ABclonal Technology BIN3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A154781000UL
Encompass_Preferred
The product of this gene is a member of the BAR domain protein family. The encoded protein is comprised solely of a BAR domain which is predicted to form coiled-coil structures and proposed to mediate dimerization, sense and induce membrane curvature, and bind small GTPases. BAR domain proteins have been implicated in endocytosis, intracellular transport, and a diverse set of other processes.
Catalog No. 50-001-06278