Learn More
Analytical Sales & Services Binary Fixed Flow Splitter Manifold 1000:1 Post Column
Catalog No. 5000806507
Encompass
Advantage Model 63 Binary Fixed Flow Splitter Manifold 1 000:1 Post Column\n\nQty/Pkg: EACH
Catalog No. 50-008-06507 Supplier Analytical Sales & Services Supplier No. 63414
Advantage Model 63 Binary Fixed Flow Splitter Manifold 1 000:1 Post Column\n\nQty/Pkg: EACH