Learn More
Analytical Sales & Services BINARY FIXED FLOW SPLITTERS
Catalog No. NC1722443
Encompass
Model 63 Post Column Binary Fixed Flow Splitters.Analytical Range: 0.1-5mL/min Incoming Flow.Binary Fixed Splitter, Ratio 3:1
Catalog No. NC1722443 Supplier Analytical Sales & Services Supplier No. 63422
Model 63 Post Column Binary Fixed Flow Splitters.Analytical Range: 0.1-5mL/min Incoming Flow.Binary Fixed Splitter, Ratio 3:1