Learn More
Abbott™ BinaxNOW™ Strep A Test
GSA_VA
Catalog No. BNX730025 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
BNX730025 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. BNX730025 Supplier Abbott™ Supplier No. 730025
Add to Cart
Add to Cart
This item is not returnable. View return policy
For the qualitative detection of Streptococcus pyogenes Group A antigen from throat swab specimens.
A membrane coated with rabbit anti-Strep A and rabbit anti-goat antibodies is combined with a rabbit anti-Strep A/goat IgG conjugate cocktail in a hinged test device.
  • Extraction Reagent On—e2M sodium nitrite with Tween 20
  • Extraction Reagent Two–0.125M acetic acid with Tween 20
  • Sterile Swabs—Designed for use in the NOW Strep A Test
  • Positive Control Swab–Heat inactivated Streptococcus pyogenes Group A dried onto swab
Order Info
Stable until the expiration dates marked on outer packaging and containers.
Compliance
CLIA Waived—87880QW; Moderately Complex—87880
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Certifications/Compliance CE Marked, FDA
Clia Complexity Waived
Detectable Analytes S. pyogenes
Sample Type Throat Swab
Type Streptococcus A Test
Includes 25 test units, 25 sterile swabs, R1 (Sodium Nitrite), R2 (Acetic Acid), positive control swab, 1 package insert
Quantity 25 Tests
Test Time 6 min.