Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott BinaxNOW™ COVID-19 Ag Card

Catalog No. BNX195000 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
40 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BNX195000 40 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BNX195000 Supplier Abbott Supplier No. 195000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Easy to use 15 min. rapid antigen test for detecting active infections of COVID-19

  • Cost-effective, high performing test designed for decentralized testing
  • Visually read results in 15 min.
    • No instrument required
  • Simple test procedure using direct nasal swab
  • Onboard extraction allows swab to be directly inserted into test card
  • Room temperature storage

Includes:

  • 40 cardboard, book-shaped hinged test cards with test strips
  • 1 x 7.5 mL extraction reagent bottle with reagent
  • 40 sterile nasal swabs
  • 1 positive control swab
    • Non-infectious recombinant SARS-CoV-2 nucleocapsid antigen
  • 1 negative control swab
  • 1 product insert
  • 1 procedure card

Notes:

  • Emergency Use Authorization (EUA) supports testing in patient care settings operating under a CLIA Certificate of Waiver, Certificate of Compliance, or Certificate of Accreditation

Specifications

Certifications/Compliance EUA
Content And Storage 2°C to 30°C
CPT Code 87811
Clia Complexity Waived
Detectable Analytes SARS-CoV-2 Nucleocapsid Protein Antigen
Detection Method Lateral flow immunoassay
Sample Type Direct Nasal Swab
Sensitivity 84.6%
Type COVID-19 Test
Disposable Single-use
For Use With (Application) Lateral Flow Immunoassay
Includes 40 cardboard, book-shaped hinged test cards with test strips, 1 x 7.5 mL extraction reagent bottle with reagent, 40 sterile nasal swabs, 1 positive control swab containing non-infectious recombinant SARS-CoV-2 nucleocapsid antigen, 1 negative control swab, 1 product insert, 1 procedure card
Material Plastic test strip with nitrocellulose
Organism Group SARS-CoV-2
Packaging Type Cardboard kit box with individually foiled wrapped test cards
Quantity 40 Tests
Specificity 0.985
Target Organism Class SARS-CoV-2
Test Time 15 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.