Learn More
Thermo Scientific™ BinaxNOW™ RSV Test Kit
Catalog No. R25430 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
42/Pk.
Catalog No. Quantity
R25430 42/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R25430 Supplier Thermo Scientific™ Supplier No. R25430
Add to Cart
Add to Cart
This item is not returnable. View return policy
Detect RSV in 15 minutes with BinaxNOW RSVRapid, a one-step lateral flow test.
Includes:
Test devices, pipets, control swabs and control reagents
TRUSTED_SUSTAINABILITY
Specifications
Description NOW RSV (Respiratory Syncytial Virus) Test Kit
Includes Test devices, pipets, control swabs and control reagents
Quantity 42/Pk.