Learn More
Apexbio Technology LLC Bindarit 130641-38-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062045
Encompass_Preferred
Bindarit (CAS 130641-38-2) also known as AF-2838 is a synthetic small-molecule inhibitor targeting the CC chemokines MCP-1 (CCL2) MCP-2 (CCL8) and MCP-3 (CCL7) By suppressing the transcription and production of MCP proteins Bindarit reduces monocyte chemoattraction and inflammation In vitro data indicate that Bindarit inhibits LPS-induced MCP-1 synthesis in monocytes with an IC50 around 172 M and selectively decreases MCP expression without impacting unrelated cytokines such as IL-6 or IL-8 Animal studies demonstrate its potential to reduce inflammatory cell infiltration vascular smooth muscle cell proliferation and neointimal hyperplasia in rodent injury models highlighting its value for inflammation and cardiovascular disease research
Catalog No. 50-000-62045 Supplier Apexbio Technology LLC Supplier No. B21565.1
Bindarit (CAS 130641-38-2) also known as AF-2838 is a synthetic small-molecule inhibitor targeting the CC chemokines MCP-1 (CCL2) MCP-2 (CCL8) and MCP-3 (CCL7) By suppressing the transcription and production of MCP proteins Bindarit reduces monocyte chemoattraction and inflammation In vitro data indicate that Bindarit inhibits LPS-induced MCP-1 synthesis in monocytes with an IC50 around 172 M and selectively decreases MCP expression without impacting unrelated cytokines such as IL-6 or IL-8 Animal studies demonstrate its potential to reduce inflammatory cell infiltration vascular smooth muscle cell proliferation and neointimal hyperplasia in rodent injury models highlighting its value for inflammation and cardiovascular disease research