Learn More
Cayman Chemical BINDARIT 25MG
SDP
Supplier:  Cayman Chemical 1147925
Encompass_Preferred
Bindarit is an inhibitor of monocyte chemoattractant protein (MCP) production that is selective for MCP-1/CCL2, MPC-3/CCL7, and MCP-2/CCL8 over other chemokines. CAS# 130641-38-2
Catalog No. NC1612372