Learn More
Apexbio Technology LLC Bindarit, 50mg. Cas: 130641-38-2 MFCD: MFCD00866723. CCL2, CCL7 and CCL8 inhibitor
SDP
Supplier:  Apexbio Technology LLC B215650
Encompass_Preferred
Bindarit(AF-2838), an anti-inflammatory agent, is a selective inhibitor againstmonocyte chemotactic proteins MCP-1/CCL2, MCP-3/CCL7 and MCP-2/CCL8[1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5319