Learn More
TARGETMOL CHEMICALS INC BINDARIT 5MG
SDP
Catalog No. 502576522
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bindarit (AF2838) displays specific inhibition against monocyte chemotactic proteins MCP-1/CCL2 MCP-3/CCL7 and MCP-2/CCL8. purity: 98%
Catalog No. 50-257-6522 Supplier TARGETMOL CHEMICALS INC Supplier No. T64135MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bindarit (AF2838) displays specific inhibition against monocyte chemotactic proteins MCP-1/CCL2 MCP-3/CCL7 and MCP-2/CCL8. purity: 98%