Learn More
Abcam Bindarit, CCL2, CCL7 and CCL8 inhibitor, 50MG
Catalog No. 502706123
Encompass_Preferred
MW 324.4 Da, Purity >99%. CCL2, CCL7 and CCL8 inhibitor. Anti-inflammatory agent. Shows antiproliferative, antimetastatic and antitumor effects in vivo. Prevents neuroinflammation in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6123 Supplier Abcam Supplier No. AB14329250MG
MW 324.4 Da, Purity >99%. CCL2, CCL7 and CCL8 inhibitor. Anti-inflammatory agent. Shows antiproliferative, antimetastatic and antitumor effects in vivo. Prevents neuroinflammation in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement