Learn More
Research Products International Corp Father Time's Binders, Medium, 3 cm Wide, 12 mm Opening, 40 per Case
SDP
Catalog No. 50212200
Encompass
Father Time's Binders are made of corrosion proof stainless steel for years of service. They eliminate rust stains and other problems associated with office style binders used in electrophoresis. Father Time's binders will not rust in acid or urea solutions and are corrosion tested in acid. Binders maintain firm tension when clamping glass plates to electrophoresis apparatus. Handles can be removed for permanent binding.
Catalog No. 50-212-200 Supplier Research Products International Corp Supplier No. 141074
Father Time's Binders are made of corrosion proof stainless steel for years of service. They eliminate rust stains and other problems associated with office style binders used in electrophoresis. Father Time's binders will not rust in acid or urea solutions and are corrosion tested in acid. Binders maintain firm tension when clamping glass plates to electrophoresis apparatus. Handles can be removed for permanent binding.