Learn More
Sigma Aldrich Fine Chemicals Biosciences Binder drying oven ED seri1EA
Catalog No. 502908950
Encompass
Natural convection ovens for all sterilization and drying tasks. It is also optimally suited to precise warm storage. Thanks to their temperature accurancies and efficiency the drying ovens and heating chambers in the ED series are very economical.APT.Line Preheating chamber technologyDigital temperature setting with ramp functionTemperature safety device CI.22 chrome plated shelvesAdjustable ventilation by means of rear exhaust duct diameter 50 mm with ventilation flap and front ventilation slide
Catalog No. 50-290-8950 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6833291EA
Natural convection ovens for all sterilization and drying tasks. It is also optimally suited to precise warm storage. Thanks to their temperature accurancies and efficiency the drying ovens and heating chambers in the ED series are very economical.APT.Line Preheating chamber technologyDigital temperature setting with ramp functionTemperature safety device CI.22 chrome plated shelvesAdjustable ventilation by means of rear exhaust duct diameter 50 mm with ventilation flap and front ventilation slide