Learn More
Thermo Scientific™ Binding Buffer for GeneJET Gel Extraction Kit
Catalog No. R1331
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 mL
Catalog No. Quantity
R1331 150 mL
1 options
Catalog No. R1331 Supplier Thermo Scientific™ Supplier No. R1331
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart
Thermo Scientific GeneJET Binding Buffer is a component of the GeneJET Gel Extraction Kit and may be purchased separately.
Thermo Scientific GeneJET Binding Buffer is a component of the GeneJET Gel Extraction Kit (Cat. No. K0691, K0692) and may be purchased separately.
Thermo Scientific GeneJET Binding Buffer is a component of the GeneJET Gel Extraction Kit (Cat. No. K0691, K0692) and may be purchased separately.
Related Products
  • GeneJET NGS Cleanup Kit (Cat. No. K0851)
  • Binding Buffer for GeneJET NGS Cleanup Kit (R2811)
  • Prewash Buffer for GeneJET NGS Cleanup Kit (R2821)
  • Wash Buffer for GeneJET NGS Cleanup Kit (R2831)
  • GeneJET Plasmid Miniprep Kit (K0502)
  • RNase A (R1253)
  • Elution Buffer for GeneJET Plasmid Miniprep Kit (R1263)
  • Lysis Solution for GeneJET Plasmid Miniprep Kit (R1223)
  • Resuspension Solution for GeneJET Plasmid Miniprep Kit (R1213)
  • Neutralization Solution for GeneJET Plasmid Miniprep Kit (R1233)
  • Wash Solution for GeneJET Plasmid Miniprep Kit (R1243)
TRUSTED_SUSTAINABILITY
Specifications
Product Line GeneJET
Product Type Binding Buffer
Quantity 150 mL
Size 150 mL
Do you offer the binding buffer included in the GeneJET PCR Purification Kit as a stand-alone product?
Yes, we offer Binding Buffer for GeneJET Gel Extraction Kit as a stand-alone product: Cat. No. R1331.
For Research Use Only. Not for use in diagnostic procedures.