Learn More
Medchemexpress LLC HY-15202 100mg Medchemexpress, Binimetinib CAS:606143-89-9 Purity:>98%
SDP
Catalog No. 502029096
Encompass_Preferred
Medchemexpress, HY-15202 100mg Binimetinib CAS:606143-89-9 Binimetinib (MEK162) is an oral and selective MEK1/2 inhibitor. Binimetinib (MEK162) inhibits MEK with an IC50 of 12 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9096 Supplier Medchemexpress LLC Supplier No. HY15202100MG
Medchemexpress, HY-15202 100mg Binimetinib CAS:606143-89-9 Binimetinib (MEK162) is an oral and selective MEK1/2 inhibitor. Binimetinib (MEK162) inhibits MEK with an IC50 of 12 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.