Learn More
Medchemexpress LLC Binimetinib | 606143-89-9 | C17H15BrF2N4O3 | 1 G
SDP
Catalog No. 5000373579
Encompass_Preferred
Binimetinib is an oral and selective MEK1/2 inhibitor that inhibits MEK with an IC50 of 12 nM. This compound is primarily utilized in research for its role in the MAPK/ERK pathway and autophagy.
  • Potent and selective MEK1/2 inhibitor with an IC50 of 12 nM.
  • Targets MEK and autophagy, impacting the MAPK/ERK pathway.
  • Blocks basal ERK phosphorylation in HRAS mutant cell lines.
  • Demonstrates synergistic effects in combination treatments.
  • Reverses resistance in RSK-expressing cells when combined with PI3K inhibition.
  • Reduces disease severity and joint destruction in animal models of arthritis.
  • Enhances efficacy when used in combination therapies for inflammation and tumor growth.
Catalog No. 50-003-73579 Supplier Medchemexpress LLC Supplier No. HY152021G
Binimetinib is an oral and selective MEK1/2 inhibitor that inhibits MEK with an IC50 of 12 nM. This compound is primarily utilized in research for its role in the MAPK/ERK pathway and autophagy.
  • Potent and selective MEK1/2 inhibitor with an IC50 of 12 nM.
  • Targets MEK and autophagy, impacting the MAPK/ERK pathway.
  • Blocks basal ERK phosphorylation in HRAS mutant cell lines.
  • Demonstrates synergistic effects in combination treatments.
  • Reverses resistance in RSK-expressing cells when combined with PI3K inhibition.
  • Reduces disease severity and joint destruction in animal models of arthritis.
  • Enhances efficacy when used in combination therapies for inflammation and tumor growth.