Learn More
Abcam Binimetinib, MEK1/2 inhibitor, 25MG
Catalog No. 502674965
Encompass_Preferred
MW 441.2 Da, Purity >99%. Potent and selective allosteric inhibitor of MEK1/2. Orally bioavailable. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-4965 Supplier Abcam Supplier No. AB25444925MG
MW 441.2 Da, Purity >99%. Potent and selective allosteric inhibitor of MEK1/2. Orally bioavailable. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement