Learn More
World Precision Instrument Binocular Loupe 2.5x, 34cm
SDP
Catalog No. 50379931
Encompass
Fully adjustable viewing angle. Compatible with most types of glass frames. The clip on loupes are a  quick and easy solution for professionals who prefer to wear their own prescription glasses. Our model is a flip-up design allowing you to view an area without removing them. These loupes are not as stables as the ones fixed onto the frame, and they may require  constant readjustments,  since movements may change the position of the loupes.
This item is not returnable. View return policy
Catalog No. 50-379-931 Supplier World Precision Instrument Supplier No. 504037
Fully adjustable viewing angle. Compatible with most types of glass frames. The clip on loupes are a  quick and easy solution for professionals who prefer to wear their own prescription glasses. Our model is a flip-up design allowing you to view an area without removing them. These loupes are not as stables as the ones fixed onto the frame, and they may require  constant readjustments,  since movements may change the position of the loupes.