Learn More
Strem, An Ascensus Company 100mg. (11bR)-2,6-Bis[2-methoxyphenyl-5-(tert-butyl)]-4-hydroxy-4-oxide-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin, 98%, (99% ee).
SDP
Supplier:  Strem, An Ascensus Company 158524100MG
Encompass
100mg. (11bR)-2,6-Bis[2-methoxyphenyl-5-(tert-butyl)]-4-hydroxy-4-oxide-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin, 98%, (99% ee). . Molecular Weight: 672.7. Molecular Formula: C42H41O6P. Color/form: off white to light yellow pwdr. Strem# 15-8524. http://www.strem.com/catalog/v/15-8524/
Catalog No. 50-206-5438