Learn More
Cayman Chemical BInucleIn 2 10mg
SDP
Catalog No. 502993849
Encompass_Preferred
An inhibitor of Drosophila Aurora B kinase (Ki = 0.36 µM); specific for Drosophila B kinase, with minimal inhibition of human or X. laevis Aurora B kinases at concentrations up to 100 µM; induces mitotic and cytokinesis defects in Drosophila Kc167 cells; prevents Drosophila S2 cells from assembling a contractile ring during cell division when used at a concentration of 40 µM
Catalog No. 50-299-3849 Supplier Cayman Chemical Supplier No. 2336710 MG
An inhibitor of Drosophila Aurora B kinase (Ki = 0.36 µM); specific for Drosophila B kinase, with minimal inhibition of human or X. laevis Aurora B kinases at concentrations up to 100 µM; induces mitotic and cytokinesis defects in Drosophila Kc167 cells; prevents Drosophila S2 cells from assembling a contractile ring during cell division when used at a concentration of 40 µM