Learn More
Medchemexpress LLC BIO-11006 acetate | 00-00-0 | 99.9% | 1110.22 | C48H79N13O17 | 100 MG
SDP
Catalog No. 5000243451
Encompass_Preferred
BIO-11006 acetate is the acetate salt of BIO-11006, an analog of the MANS peptide that inhibits MARCKS (myristoylated alanine-rich C kinase substrate). Supplied for laboratory research use only, the peptide salt is provided with characterization data to support in vitro studies of MARCKS-related signaling and membrane interactions.
  • High purity suitable for research (>99.9%).
  • Acetate salt form for improved handling and stability.
  • Relevant for in vitro studies of MARCKS-mediated signaling and peptide-membrane interactions.
  • Available in laboratory-scale pack sizes to support assay development.
  • Characterized molecular weight and formula provided for experimental planning.
Catalog No. 50-002-43451 Supplier Medchemexpress LLC Supplier No. HY106377A100MG
BIO-11006 acetate is the acetate salt of BIO-11006, an analog of the MANS peptide that inhibits MARCKS (myristoylated alanine-rich C kinase substrate). Supplied for laboratory research use only, the peptide salt is provided with characterization data to support in vitro studies of MARCKS-related signaling and membrane interactions.
  • High purity suitable for research (>99.9%).
  • Acetate salt form for improved handling and stability.
  • Relevant for in vitro studies of MARCKS-mediated signaling and peptide-membrane interactions.
  • Available in laboratory-scale pack sizes to support assay development.
  • Characterized molecular weight and formula provided for experimental planning.