Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bio-11006 acetate | 00-00-0 | 99.9% | 1110.22 g/mol | C48H79N13O17 | 50 MG
SDP

Catalog No. 5000242904
Encompass_Preferred

BIO-11006 acetate is the acetate salt form of a peptide analog of the MANS peptide that inhibits MARCKS (myristoylated alanine-rich C kinase substrate). It is supplied for research use and is available as lyophilized peptide vials or as a ready-to-use aqueous solution.

  • Inhibits MARCKS protein by mimicking the MANS peptide.
  • Peptide acetate salt format for improved handling and solubility.
  • Chemical formula C48H79N13O17.
  • Molecular weight 1110.22 g/mol.
  • Purity 99.92%.
  • Available in multiple pack sizes, including 50 mg and 10 mM 1 mL solution.
  • For research use only.

Catalog No. 50-002-42904 Supplier Medchemexpress LLC Supplier No. HY106377A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BIO-11006 acetate is the acetate salt form of a peptide analog of the MANS peptide that inhibits MARCKS (myristoylated alanine-rich C kinase substrate). It is supplied for research use and is available as lyophilized peptide vials or as a ready-to-use aqueous solution.

  • Inhibits MARCKS protein by mimicking the MANS peptide.
  • Peptide acetate salt format for improved handling and solubility.
  • Chemical formula C48H79N13O17.
  • Molecular weight 1110.22 g/mol.
  • Purity 99.92%.
  • Available in multiple pack sizes, including 50 mg and 10 mM 1 mL solution.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.