Learn More
Medchemexpress LLC BIO-11006 acetate 10mM 1mL | 10MM 1ML
SDP
Catalog No. 5000244352
Encompass_Preferred
BIO-11006 acetate an analog of the MANS peptide is a MARCKS (myristoylated alanine-rich C kinase substrate) inhibitor[1
Catalog No. 50-002-44352 Supplier Medchemexpress LLC Supplier No. HY106377A10MM 1ML
BIO-11006 acetate an analog of the MANS peptide is a MARCKS (myristoylated alanine-rich C kinase substrate) inhibitor[1