Learn More
Medchemexpress LLC HY-106377A 10mg Medchemexpress, BIO-11006 acetate salt CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY106377A10MG
Encompass_Preferred
Medchemexpress, HY-106377A 10mg BIO-11006 acetate salt CAS: BIO-11006 acetate salt, an analog of the MANS peptide, is a myristoylated alanine-rich C kinase substrate (MARCKS) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8341