Learn More
Enzo Life Sciences Bio-16-ddUTP (25nmol)
SDP
Supplier:  Enzo Life Sciences ENZ42813
Encompass_Preferred
Bio-16-ddUTP (Biotin-16-2’,3’-dideoxyuridine-5’-triphosphate) can be used for 3’-end labeling of single-stranded or double-stranded DNA. It is especially useful as a substrate for terminal transferase for the 3’-terminal addition of a single Bio-16-ddUMP. Biotin-labeled oligonucleotides produced by this reaction are suitable as hybridization probes. Biotin-labeled cDNA produced by this reaction can be used in nucleic acid array assays. Biotin-labeled DNA in hybridization assays can be detected by a reporter molecule linked to streptavidin, avidin, or and anti-biotin antibody. Such a complex can be detected directly, e.g. by the excitation of a fluorophore conjugated to streptavidin, or indirectly, e.g. using an enzyme conjugate that can produce an insoluble colored precipitate. Alternative name: Biotin-16-2',3'-dideoxyuridine-5'-triphosphate . 4Li. Concentration: 1mM. Purity: ≥93% (HPLC). Formulation: Liquid. Solution in water.
Catalog No. 50-201-2542