Learn More
Enzo Life Sciences Bio 17 ATP (250 nmol)
SDP
Catalog No. NC0314202
Encompass_Preferred
Alternative Name: Biotin-17-adenosine-5'-triphosphate. Formulation: Liquid. Solution in water. Purity: ≥93% (HPLC). Appearance: Clear, colorless liquid. Shipping: Shipped on Dry Ice. Long Term Storage: -20°C. Handling: Avoid freeze/thaw cycles.
Catalog No. NC0314202 Supplier Enzo Life Sciences Supplier No. ENZ42817
Alternative Name: Biotin-17-adenosine-5'-triphosphate. Formulation: Liquid. Solution in water. Purity: ≥93% (HPLC). Appearance: Clear, colorless liquid. Shipping: Shipped on Dry Ice. Long Term Storage: -20°C. Handling: Avoid freeze/thaw cycles.