Learn More
Medchemexpress LLC BIO-2007817 | 99.35% | 552.67 | 25 MG
SDP
Catalog No. 5000394295
Encompass_Preferred
BIO-2007817 is a Parkin positive allosteric modulator (PAM) that enhances the activity of wildtype Parkin. It stimulates Parkin (an E3 ligase) autoubiquitination and induces the appearance of monoubiquitinated forms of Miro1 with an EC50 of 0.17 μM.
  • Parkin positive allosteric modulator (PAM)
  • Enhances activity of wildtype Parkin
  • Stimulates Parkin (an E3 ligase) autoubiquitination
  • Induces monoubiquitinated forms of Miro1 (EC50: 0.17 μM)
  • For research use only
Catalog No. 50-003-94295 Supplier Medchemexpress LLC Supplier No. 25MGHY169329
BIO-2007817 is a Parkin positive allosteric modulator (PAM) that enhances the activity of wildtype Parkin. It stimulates Parkin (an E3 ligase) autoubiquitination and induces the appearance of monoubiquitinated forms of Miro1 with an EC50 of 0.17 μM.
  • Parkin positive allosteric modulator (PAM)
  • Enhances activity of wildtype Parkin
  • Stimulates Parkin (an E3 ligase) autoubiquitination
  • Induces monoubiquitinated forms of Miro1 (EC50: 0.17 μM)
  • For research use only