Learn More
Air Science Usa Biosafety Cabinet 48" / 1200mm Nominal Width, NSF Listed, Biological Safety Cabinet
SDP
Catalog No. NC1837398
Encompass_Preferred
Purair Bio Class II A2 Biological Safety Cabinet 48" / 1200mm Nominal Width, NSF Listed, 115V 60Hz, Includes: Adjustable Height Base Stand on Levelers, UV system, Two GFI Outlets, One Service Fixture (as-Aha-133-Ca-B)
Catalog No. NC1837398 Supplier Air Science Usa Supplier No. ASAHA133CAB
Add to Cart
Add to Cart
Request a Quote
Purair Bio Class II A2 Biological Safety Cabinet 48" / 1200mm Nominal Width, NSF Listed, 115V 60Hz, Includes: Adjustable Height Base Stand on Levelers, UV system, Two GFI Outlets, One Service Fixture (as-Aha-133-Ca-B)