Learn More
Cayman Chemical BIO-AcetoxIme 10mg
SDP
Catalog No. 502972429
Encompass_Preferred
A potent inhibitor of GSK3α/β (IC50 = 10 nM), with selectivity over Cdk5/p25, Cdk2/A, and Cdk1/B (IC50s = 2.4, 4.3, and 63 µM, respectively); suppresses the cytopathic effects of herpes viruses and reduces viral yields in human oral epithelial cells
Catalog No. 50-297-2429 Supplier Cayman Chemical Supplier No. 1632910 MG
A potent inhibitor of GSK3α/β (IC50 = 10 nM), with selectivity over Cdk5/p25, Cdk2/A, and Cdk1/B (IC50s = 2.4, 4.3, and 63 µM, respectively); suppresses the cytopathic effects of herpes viruses and reduces viral yields in human oral epithelial cells