Learn More
Selleck Chemical LLC BIO-acetoxime-5mg
SDP
Catalog No. 502390072
Encompass_Preferred
BIO-acetoxime (GSK-3 Inhibitor X) is a potent dual GSK3α/β inhibitor with IC50 of 10 nM, >240-fold selectivity over CDK5/p25, CDK2/cyclin A and CDK1/cyclin B.
Catalog No. 50-239-0072 Supplier Selleck Chemical LLC Supplier No. S79155MG
BIO-acetoxime (GSK-3 Inhibitor X) is a potent dual GSK3α/β inhibitor with IC50 of 10 nM, >240-fold selectivity over CDK5/p25, CDK2/cyclin A and CDK1/cyclin B.