Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIO-acetoxime, Tocris Bioscience™

Catalog No. 38741
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-741 1 mg
38-741-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-741 Supplier R&D Systems Supplier No. 3874/1
This item is not returnable. View return policy

Selective GSK-3α/β inhibitor

Selective GSK-3α/β inhibitor (IC50 values are 0.01, 2.4, 4.3 and 63 μM for GSK-3α/β, CDK5/p25, CDK2/cyclin A and CDK1/cyclin B respectively). Induces Wnt signaling and inhibits CD8+ T cell effector differentiation.

Chemical Identifiers

CAS 667463-85-6
Molecular Formula C18H12BrN3O3
Molecular Weight (g/mol) 398.216
InChI Key ORWSYHWTBSHFIV-BMRADRMJSA-N
Synonym bio-acetoxime, gsk-3 inhibitor x, bio-acetoxime gsk-3 inhibitor x, e,3e-6'-bromo-2'-oxo-1h,1'h-2,3'-biindolyliden-3-ylidene amino acetate
PubChem CID 77519830
IUPAC Name [[(2E)-2-(6-bromo-2-oxo-1H-indol-3-ylidene)indol-3-yl]amino] acetate
SMILES CC(=O)ONC1=C2C=CC=CC2=NC1=C3C4=C(C=C(C=C4)Br)NC3=O

Specifications

Quantity 1 mg
Formula Weight 398.21
Percent Purity >98%
Chemical Name or Material BIO-acetoxime
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.