Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bio-AMS TFA | 00-00-0 | 98.0% | 685.65 g/mol | C22H30F3N9O9S2 | 5 MG
SDP

Catalog No. 5000264832
Encompass_Preferred

Bio-AMS TFA is a bacterial biotin protein ligase inhibitor supplied as the trifluoroacetate (TFA) salt for research use. It shows selective activity against Mycobacterium tuberculosis and disrupts fatty acid and lipid biosynthesis.

  • Potent biotin protein ligase inhibition.
  • Selective activity against Mycobacterium tuberculosis.
  • Provided as the TFA salt to aid handling and solubility.
  • High purity: 98.0%.
  • Molecular weight 685.65 g/mol; formula C22H30F3N9O9S2.
  • Available in small research pack sizes, including 5 mg.
  • Suitable for biochemical and microbiology research applications.

Catalog No. 50-002-64832 Supplier Medchemexpress LLC Supplier No. HY115448A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bio-AMS TFA is a bacterial biotin protein ligase inhibitor supplied as the trifluoroacetate (TFA) salt for research use. It shows selective activity against Mycobacterium tuberculosis and disrupts fatty acid and lipid biosynthesis.

  • Potent biotin protein ligase inhibition.
  • Selective activity against Mycobacterium tuberculosis.
  • Provided as the TFA salt to aid handling and solubility.
  • High purity: 98.0%.
  • Molecular weight 685.65 g/mol; formula C22H30F3N9O9S2.
  • Available in small research pack sizes, including 5 mg.
  • Suitable for biochemical and microbiology research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.