Learn More
Bio Rad Laboratories BIO-BEADS S-X1 SUPPORT
Catalog No. 509012924
Encompass
Media, S-X; Bio-Rad; Bio-Beads; S-X1 support; Neutral; Porous styrene divinylbenzene; 1% crosslinked; For lipophilic polymers of F.W. 600-14,000; 100g; 40-80um bead
Catalog No. 50-901-2924 Supplier Bio Rad Laboratories Supplier No. 1522150
Media, S-X; Bio-Rad; Bio-Beads; S-X1 support; Neutral; Porous styrene divinylbenzene; 1% crosslinked; For lipophilic polymers of F.W. 600-14,000; 100g; 40-80um bead