Learn More
Stanbio™ Bio-Clean™ Concentrated Detergent and Deodorizer
GSA_VASDP
Catalog No. SB0602380 Shop All Stanbio Laboratory Products
Change view
Click to view available options
Volume (English):
1 gal.
Catalog No. Volume (English)
SB-0602-380 1 gal.
1 options
This item is not returnable. View return policy
Catalog No. SB-0602-380 Supplier Stanbio™ Supplier No. 0602380
Add to Cart
Add to Cart
This item is not returnable. View return policy
For cleaning laboratory instruments, glassware, porcelain, metal, rubber, and plastics
Eliminates odors on most surfaces.
  • Biodegradable and phosphate-free
  • Concentrated—use just 7mL of detergent for each gallon of water
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Application) Cleaning laboratory instruments, Glassware, surgical instruments, Porcelain, Metal, Rubber and Plastics, It also destroys odors wherever they exist on contact
Volume (English) 1 gal.