Learn More
ACROBiosystems Biotinylated Cynomolgus CD8 alpha / CD8A Protein, 200 μg, His, Avitag™, 21.3 kDa, >90% purity, Lyophilized, Storage -20°C or lower for 12 months
Catalog No. NC3495797
Encompass_Preferred
Biotinylated Cynomolgus CD8 alpha / CD8A Protein, 200 μg, His, Avitag™, 21.3 kDa, >90% purity, Lyophilized, Storage -20°C or lower for 12 months
Catalog No. NC3495797 Supplier ACROBiosystems Supplier No. CDAC82E7200UG
Biotinylated Cynomolgus CD8 alpha / CD8A Protein, 200 μg, His, Avitag™, 21.3 kDa, >90% purity, Lyophilized, Storage -20°C or lower for 12 months