Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio/Data™ Arachidonic Acid (Sodium Archidonate)
SDP

Catalog No. 22515230
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-515-230 3 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-515-230 Supplier Bio/Data™ Supplier No. 101297
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for routine platelet aggregation studies to differentiate aspirin-like defects and storage pool disease

  • Evaluate inhibitory effect of aspirin on platelet aggregation
  • Lyophilized preparation of sodium arachidonate
  • Lyophilization enables long term storage and stability
  • Screen or confirm ASA use
  • Working concentration: 5mg/mL (500μg/mL)
  • Final concentration: 0.05g/mL (50μg/mL)

Specifications

Concentration 5 mg/mL
Type Arachidonic Acid
For Use With (Application) Routine platelet aggregation studies for in-vitro use, differential diagnosis of Aspirin-like release defects and storage pool disease, to evaluate the inhibitory effect of Aspirin on platelet aggregation
Quantity 3 x 0.5 mL
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.