Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Data Collagen
SDP

Catalog No. 22515236
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-515-236 3 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-515-236 Supplier Bio Data Corporation Supplier No. 101562
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Used for evaluation of platelet dysfunction or activation

  • Lyophilized preparation of soluble calf skin collagen (Type I)
  • Lyophilization enables long term storage and stability
  • Used for measuring lag phase
  • Working concentration: 1.9mg/mL (1900μg/mL)
  • Final concentration: 0.19mg/mL (190μg/mL)
  • Quantity: 3 x 0.5mL

Specifications

Concentration 1.9 mg/mL
For Use With (Application) Lag Phase Measurement, Platelet Aggregation
pH Range <7
Quantity 3 x 0.5 mL
Source Soluble Calf Skin, Type I (Fibrillar) Collagen
Product Type Collagen
Content And Storage 2°C to 8°C
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.