Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio/Data™ Epinephrine (Adrenaline)
SDP

Catalog No. 22515245
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-515-245 3 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-515-245 Supplier Bio/Data™ Supplier No. 101311
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for evaluation of platelet dysfunction or activation

  • Lyophilized preparation of adrenalin
  • Lyophilization enables long term storage and stability
  • Working concentration: 1x10-3 M (1000μM)
  • Final concentration 1x10-4 M (100μM)

Specifications

Concentration 1 x 10-3M
Type Epinephrine
For Use With (Application) For use in routine platelet aggregation studies for the evaluation of platelet dysfunction or platelet activation
Quantity 3 x 0.5 mL
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.