Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio/Data™ PAR Pak
SDP

Catalog No. 22515254
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-515-254 2 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-515-254 Supplier Bio/Data™ Supplier No. 101310
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For routine platelet aggregation studies to evaluate platelet dysfunction or activation

  • Contains lyophilized ADP, Epinephrine, and Type I Collagen (two vials of each reagent)
  • Lyophilization enables long term storage and stability

Specifications

Type Para-Pak™ II
For Use With (Application) For use in routine platelet aggregation studies for the evaluation of platelet dysfunction or platelet activation
Includes ADP (Adenosine-5'-diphosphate), Epinephrine and soluble calf skin collagen (Type I)
Quantity 2 x 0.5 mL
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.