Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio/Data™ Ristocetin Reagent
SDP

Catalog No. 22515226
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.015 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-515-226 0.015 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-515-226 Supplier Bio/Data™ Supplier No. 100970
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Detection of von Willebrand Syndrome and quantification of von Willebrand factor

  • Lyophilized Ristocetin A sulfate
  • Isolated from Nocardia Lurida
  • In excess of 90% Ristocetin A
  • Confirms diagnosis of type IIb vWD
  • Identify Bernard-Soulier syndrome
  • Used for ristocetin induced platelet aggregation

Specifications

Concentration 1 to 1.5 mg/mL
Type Ristocetin A Sulfate
Quantity 0.015 g
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.