Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio/Data™ Sodium Citrate
SDP

Catalog No. 22515269
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
237 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-515-269 237 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-515-269 Supplier Bio/Data™ Supplier No. 100994
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Works as an anticoagulant for platelet aggregation blood sample collection

  • Formulated at 0.11M as per International Committee of Thrombosis & Hemostasis, Subcommittee on Platelets
  • Whole blood to sodium citrate ratio 9 to 1

Specifications

Concentration 0.11M
Type Sodium Citrate
For Use With (Application) For platelet aggregation Blood sample collection and as anticoagulant for coagulation Blood sample collection
Quantity 237 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.