Learn More
Bio Rad Laboratories BIO-DOT APPARATUS
Catalog No. 509010018
Encompass
Apparatus; Bio-Rad; Bio-Dot; Autoclavable; Resistant to alcohol, harsh acids, bases; Includes Bio-Dot sample template, vacuum manifold base, gasket support plate, gasket; 3mm dia. well size; 50-600uL sample vol.; 15L x 13W x 6cm H
Catalog No. 50-901-0018 Supplier Bio Rad Laboratories Supplier No. 1706545
Apparatus; Bio-Rad; Bio-Dot; Autoclavable; Resistant to alcohol, harsh acids, bases; Includes Bio-Dot sample template, vacuum manifold base, gasket support plate, gasket; 3mm dia. well size; 50-600uL sample vol.; 15L x 13W x 6cm H