Learn More
Sanisure Inc Bio-Ease+™ HD Pinch Clamps
Catalog No. NC3786304
Encompass_Preferred
The SaniSure Bio-Ease™ HD clamp is designed for critical fluid applications where regulatory compliance, seal integrity, and repeatability are of the utmost importance. Its is designed to handle higher pressures and be more secure than a standard pinch clamp. The material of construction is a USP Class VI white glass-filled nylon which can be autoclaved or gamma irradiated in cleanroom packaged kits. Bio-Ease HD clamps can also be used with reinforced hose such as our Pharma-Clear braided silicone.
Catalog No. NC3786304 Supplier Sanisure Inc Supplier No. BFS101MOQ 10
The SaniSure Bio-Ease™ HD clamp is designed for critical fluid applications where regulatory compliance, seal integrity, and repeatability are of the utmost importance. Its is designed to handle higher pressures and be more secure than a standard pinch clamp. The material of construction is a USP Class VI white glass-filled nylon which can be autoclaved or gamma irradiated in cleanroom packaged kits. Bio-Ease HD clamps can also be used with reinforced hose such as our Pharma-Clear braided silicone.