Learn More
Sanisure Inc Bio-Ease+™ Sanitary Clamps 2"
Catalog No. NC2219156
Encompass_Preferred
SaniSure Bio-Ease+™ is a patented non-metallic sanitary clamp designed to meet the needs of various single-use applications in the pharmaceutical and bio-processing industries. Manufactured using a glass-reinforced, USP Class VI Nylon and designed to be used as a component for single-use transfer lines and a wide range of bio-process systems, these plastic clamps are much lighter than their stainless steel counterparts. SaniSure stocks a comprehensive inventory of sizes from Mini to 4″. Additionally, these clamps fit ASME BPE Tri-Clamp connections, ensuring compatibility with industry-standard equipment.
This item is not returnable. View return policy
Catalog No. NC2219156 Supplier Sanisure Inc Supplier No. 13MHHS20001
Please call customer service for current pricing. 1-800-766-7000
SaniSure Bio-Ease+™ is a patented non-metallic sanitary clamp designed to meet the needs of various single-use applications in the pharmaceutical and bio-processing industries. Manufactured using a glass-reinforced, USP Class VI Nylon and designed to be used as a component for single-use transfer lines and a wide range of bio-process systems, these plastic clamps are much lighter than their stainless steel counterparts. SaniSure stocks a comprehensive inventory of sizes from Mini to 4″. Additionally, these clamps fit ASME BPE Tri-Clamp connections, ensuring compatibility with industry-standard equipment.