Learn More
Research Products International Corp Bio-Filtrators, Sterile Vacuum Filter, PVDF Membrane, 0.22µm, 1 Liter Capacity, 12 per Case
SDP
Supplier:  Research Products International Corp 195475
Encompass
Sterile Vacuum Bio-Filtrators have a built in 50mm diameter filter with a pore size of 0.22µm. Ideal for use in filtering biological fluids, culture media and other aqueous solutions. Item has a wide bottle opening for easy access and pouring the product. The funnel and bottle are manufactured from optically clear polystyrene. The filter adapter is manufactured from 100% virgin polypropylene.
Catalog No. 50-197-7315